Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzethonium chloride
Benzethonium chloride 0.03 N, AVS TITRINORM volumetric solution
Benzethonium chloride 0.03 N, AVS TITRINORM volumetric solution
89577.320
:
VWR Chemicals
:
Benzethonium chloride 0.03 N, AVS TITRINORM volumetric solution
89577.320
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
2,5 L
Seq:
5096
Titer:
0.03000 ±0.5%
:
:
448.09 g/mol
:
121-54-0
:
C₂₇H₄₂ClNO₂
:
MFCD00011742