Multi-element standard solution 6 for ICP, TraceCERT®
Multi-element standard for 23 elements, 100 mL
Supplier: Merck
Multi-element standard solution 6 for ICP, TraceCERT®
Catalog # 43843-100ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
This standard solution is intended for use as a certified reference material for ICP/AAS.
Multi element standards for Al, Sb, Ba, Pb, B, Ca, Cd, Cr, Co, Fe, K, Cu, Li, Mg, Mn, Mo, Na, Ni, P, Si, Ti, V and Zn at 100 mg/ml in 5% nitric acid and ≤0,1% hydrofluoric acid.
Certifications: Accredited to ISO 17034 and ISO/IEC 17025.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100 mL
- Description:Multi-element standard for 23 elements
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



