Pump tubing, Tygon® MHLL 2765-175
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Longest flex life of any organic solvent resistant tubing.
- Plasticizer free
- Virtually no extractables
Especially designed for MEK and other aggressive solvents.
Temperature resistance: −70 to +74 °C
Hardness (Shore durometer A): 64
Certifications: Complies with US Pharmacopoea, Class VI, US Pharmacopoea, Klasse VI, FDA 21CFR 177.2600 criterias.
Frequently Bought Together