Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Penetration probe

Penetration probe PT100 with 2 m fixed ribbon cable, −85…+150 °C, Class A, 5×60 mm
  620-1971
 :   TESTO
Penetration probe
  620-1971
 :  TESTO
Penetration probe
Penetration probe

Penetration probe PT100 with 2 m fixed ribbon cable, −85…+150 °C, Class A, 5×60 mm

 
120,00 €
   
1 * 1 ST
 
 
  • Range:
    −85…+150 °C
  • Description:
    Penetration probe PT100 with 2 m fixed ribbon cable
  • Accuracy:
    Class A
  • Ø×L:
    5×60 mm