Turbidity calibration standards, formazin, Supelco®
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Laboratory bottles, narrow neck, with thread, DURAN®
: DWK Life Sciences
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
: VWR Chemicals
Venting filters, Vacushield™
: Cytiva (Formerly Pall Lab)
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
VWR®, Turbidity standards, formazin
: VWR Chemicals