Orders LinkContactUsLinkComponent
Percoll®, Siam
Percoll®, Siam
Percoll®, Siam
Certificates
4 Options Available Below
Product Details & Documents
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
  • Non toxic
  • Ideal for use with biological materials
Documents
Product Family Options
Product Information
  • Availability
    Price
  • Percoll®
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 060,00 €
    1 060,00 €
    1 * 1 L
       1 060,00 €
  • P1644-25ML
    Percoll®
    Percoll®
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 69,40 €
    69,40 €
    1 * 25 mL
       69,40 €
  • P1644-100ML
    Percoll®
    Percoll®
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 198,00 €
    198,00 €
    1 * 100 mL
       198,00 €
  • P1644-500ML
    Percoll®
    Percoll®
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 604,00 €
    604,00 €
    1 * 500 mL
       604,00 €