Order Entry
Belgium
ContactUsLinkComponent
Ammonia solution 25%, LiChropur™ for LC-MS, Supelco®
Corrosive
Irritant
Environment
Ammonia solution 25%, LiChropur™ for LC-MS, Supelco®
Catalog # 5.33003.0050
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Environment
Ammonia solution 25%, LiChropur™ for LC-MS, Supelco®
Catalog # 5.33003.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Density (20 °C):
    0,903 g/cm³
  • Explosion limit (V):
    15,4 – 33,6%
  • Vapor pressure (20 °C):
    483 hPa
  • Assay:
    25%
  • Storage:
    Store at +15 °C – +25 °C

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:37.7 °C
Melting Pt:–57,5 °C
Density:0.903 g/cm³ (20 °C)
UN:2672
ADR:8,III
 :M-Clarity™ quality level = MQ100
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650