Orders LinkContactUsLinkComponent
Extractors for solids
Extractors for solids
Supplier:   Lenz Laborglas
Extractors for solids
Supplier:  Lenz Laborglas
Certificates
Extractors for solids
Extractors for solids
3 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of solids (warm).
The complete apparatuses are consisting of:
Extractor head with glass thimbles for solids, Dimroth condenser, with thread GL 14, round bottom flask.
Documents
Product Family Options
Product Information
  • Extraction cap.
    Flask capacity
    Socket size
    Availability
    Price
  • 70 ml
    250 ml
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • 100 ml
    250 ml
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 268,00 €
    268,00 €
    1 * 1 ST
       268,00 €
  • 250 ml
    500 ml
    60/46 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 371,00 €
    371,00 €
    1 * 1 ST
       371,00 €