ContactUsLinkComponent
Intubation cannula
Intubation cannula
Catalog # HARV73-2855
Supplier:  HARVARD APPARATUS
undefined
Intubation cannula
Catalog # HARV73-2855
Supplier:  HARVARD APPARATUS
Specifications
  • Ø×L:
    2,5×50 mm
  • Description:
    Intubation cannula for rats and Guinea pigs
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This intubation cannula is used in chronic studies where the animal needs to be intubated.
  • Outside diameter must fit to the animal’s trachea
  • Luer-adapter for use with any Luer connection
  • Available in several sizes for various species