Order Entry
Belgium
ContactUsLinkComponent
Calcium sulphate, Drierite™ 6 mesh, with indicator
Irritant
Health
Environment
Calcium sulphate, Drierite™ 6 mesh, with indicator
Certificates
undefined
Irritant
Health
Environment
Calcium sulphate, Drierite™ 6 mesh, with indicator

Specification Test Results

  • Appearance:
    Blue Irregular granules
  • Water:
    ≤0.3 %
  • Particle size:
    6 mesh
  • Composition:
    ≥98 % (CaSO4) Typical

Specification Test Results

Frequently Bought Together