ContactUsLinkComponent
Tubes with screw cap, PP
  SARS60.540.016
 :  SARSTEDT
Tubes with screw cap, PP
  SARS60.540.016
 :  SARSTEDT
  • Ext.Ø:
    16,5 mm
  • Capacity:
    13 ml
  • Length:
    101 mm
  • Environmentally Preferable:
  • Pk:
    500
PP, translucent, with assembled integral O-ring screw cap.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sterile
Round bottom, with attached screw cap.