Permanent marker pen, PLUS
Marker pen, Black
Catalog # KLIN98307-R
Supplier: KLINIPATH
Jump to:
Product Details & Documents
This black permanent marker pen is ideal for marking slides, embedding cassettes or dotting cells.
- Solvent resistant
- Odourless
- Fast drying
Specifications
- Colour:Black
- Description:Marker pen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



