ContactUsLinkComponent
Drill handpiece, Aesculap®
Drill handpiece, Aesculap®
Catalog # AESCGB169R
Supplier:  Aesculap
Drill handpiece, Aesculap®
Catalog # AESCGB169R
Supplier:  Aesculap
Specifications
  • Description:
    Universal drill hand piece, for exchangeable chucks, 900 rpm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Drill handpiece for exchangeable chucks. Features power and stability during skull perforations.
  • Proven versatility at low speed
  • Longer and slimmer design for additional visibility
Ordering information: Motor, control unit, spray nozzle and cables need to be ordered separately.
Delivery: Includes a ring spanner.