ContactUsLinkComponent
Anaerobic culture tube, Hungate type
Anaerobic culture tube, Hungate type
Catalog # BELC2047-16125
Supplier:  BELLCO GLASS
undefined
Anaerobic culture tube, Hungate type
Catalog # BELC2047-16125
Supplier:  BELLCO GLASS
Specifications
  • Ø:
    16 mm
  • Height:
    125 mm
  • Pk:
    250
Specifications
About this item
These tubes made of type 1 class B borosilicate glass for growing and maintaining anaerobic bacteria, culture condition.
  • Very good temperature resistance
  • Very good chemical resistance