GL transition fittings, BOLA
Fitting made of PTFE, white.
- Temperature resistance from –200 to +250 °C
- Universal chemical resistance
- FDA conform
One side with thread GL 14 for connecting hard-walled tubing and tubes by means of BOLA laboratory screw joints. Other side with female thread UNF 1/4" 28G for connecting flanged tubing with O. D. 1,6 and 3,2 mm by means of tube end fittings.
Frequently Bought Together