Orders LinkContactUsLinkComponent
Jars, without Cap
Jars, without Cap
Jars, without Cap
Jars, without Cap
Jars, without Cap
These slip lid jars are made of white PP.
  • Very good chemical resistance
  • Very good temperature resistance
Ordering information: Caps need to be ordered seperately.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
  • Type
    Capacity
    Ext.Ø
    Neck I-Ø
    Height
  • PP
    25 ml
    27,6 mm
    25,0 mm
    52,2 mm
    1 * 2.000 ST
    1 * 2.000 ST - 536,00 €
    536,00 €
    1 * 2.000 ST
       536,00 €
  • PP
    50 ml
    37,7 mm
    36,0 mm
    73,8 mm
    1 * 900 ST
    1 * 900 ST - 403,00 €
    403,00 €
    1 * 900 ST
       403,00 €
  • PP
    100 ml
    47,4 mm
    46,0 mm
    86,5 mm
    1 * 500 ST
    1 * 500 ST - 257,00 €
    257,00 €
    1 * 500 ST
       257,00 €
  • PP
    250 ml
    83,4 mm
    1 * 200 ST
    1 * 200 ST
    New Product
    New Product
  • PP
    325 ml
    69,0 mm
    66,0 mm
    113, 0 mm
    1 * 180 ST
    1 * 180 ST - 267,00 €
    267,00 €
    1 * 180 ST
       267,00 €
  • PP
    350 ml
    95,2 mm
    90,0 mm
    68,0 mm
    1 * 172 ST
    1 * 172 ST - 248,00 €
    248,00 €
    1 * 172 ST
       248,00 €
  • PP
    500 ml
    94,8 mm
    91,0 mm
    85,0 mm
    1 * 123 ST
    1 * 123 ST - 263,00 €
    263,00 €
    1 * 123 ST
       263,00 €
Caps
  •  
    Colour
    Material
    For
  • White
    HDPE
    Jar, 50 ml
    1 * 25 ST
    1 * 25 ST - 5,00 €
    5,00 €
    1 * 25 ST
       5,00 €
  •  
    Colour
    Material
    For
  • White
    HDPE
    Jar, 325 ml
    1 * 25 ST
    1 * 25 ST - 12,90 €
    12,90 €
    1 * 25 ST
       12,90 €
  •  
    Colour
    Material
    For
  • White
    HDPE
    Jar, 100 ml
    1 * 25 ST
    1 * 25 ST - 6,90 €
    6,90 €
    1 * 25 ST
       6,90 €
  •  
    Colour
    Material
    For
  • White
    HDPE
    Jar, 500 ml
    1 * 25 ST
    1 * 25 ST - 27,50 €
    27,50 €
    1 * 25 ST
       27,50 €