Cleaning coupons
Supplier: Sampling Systems
Jump to:
About this item
These cleaning validation coupons are available in all popular materials and various sizes.
6 Options Available Below
Product Details & Documents
These cleaning validation coupons are available in all popular materials and various sizes.
Product Family Options
Product Information
- L×WAvailabilityPrice
- SAMP8644S6-50BCleaning coupon, 316 stainless steel (bright polish)Cleaning coupon, 316 stainless steel (bright polish)50×50 mmLog in for availability1 * 1 ST238,00 €
- SAMP8644T-50Cleaning coupon, PTFE
- SAMP8644S4-100BCleaning coupon, 304 stainless steel (bright polish)Cleaning coupon, 304 stainless steel (bright polish)100×100 mmLog in for availability1 * 1 ST275,00 €
- SAMP8644S6-100BCleaning coupon, 316 stainless steel (bright polish)Cleaning coupon, 316 stainless steel (bright polish)100×100 mmLog in for availability1 * 1 ST301,00 €
- SAMP8644S6-100SCleaning coupon, 316 stainless steel (satin finish)Cleaning coupon, 316 stainless steel (satin finish)100×100 mmLog in for availability1 * 1 ST270,00 €
- SAMP8644T-100Cleaning coupon, PTFE