Orders LinkContactUsLinkComponent
Hydrogen generators
Hydrogen generators
Supplier:   DOMNICK HUNTER
Hydrogen generators
Supplier:  DOMNICK HUNTER
Certificates
Hydrogen generators
Hydrogen generators
1 Options Available Below
Product Details & Documents
Parker Domnick Hunter hydrogen generators produce a continuous flow of pure hydrogen from deionised water. These hydrogen generators provide a safe and hassle-free alternative to high pressure gas cylinders. Hydrogen is only generated on demand at low pressure and the volume of stored gas is minimal.
  • Automatic hydrogen leak detection
  • Patented seven-stage explosion protection system
  • Continuous monitoring of 'vital parameters'
  • Easy to use controller, high visibility/audible alarms
  • Large capacity water tank/optional auto water fill
  • Optional remote control and monitoring
These generators are ideal for supplying fuel gas to all known combustion detectors used routinely in GC and can also be used to supply hydrogen for GC carrier gas applications and for ELCD reaction gas.
To ensure optimum performance and longevity of these hydrogen generators, preventative maintenance kits are recommended.
Contact Avantor for your installation and service requirements.
Contact VWR for your installation and service requirements.
Product Family Options
Ultra-high purity: >99,99995% H₂; 0,69 - 6,89 bar for GC and GC-MS carrier gas applications
Product Information
  • Type
    Flow rate
    W×D×H
    Availability
    Price
  • 20H-MD
    160 ml/min
    342×470×456 mm
    Discontinued
  • 110H-MDCARRIER
    1100 ml/min
    342×470×456 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST