Order Entry
Belgium
Orders LinkContactUsLinkComponent
Software for data loggers

Software for data loggers

Windows software, GSOFT 40K
Catalog # 662-5511
Supplier:   Greisinger
Software for data loggers
Catalog # 662-5511
Supplier:  Greisinger
Software for data loggers

Windows software, GSOFT 40K

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

This windows-software is suitable for EASYLog and T-Logg with logger.

  • Automated retrieval and archiving
  • Data export of series of measurements as a CSV file
  • Print out the data as a table and/or trace
  • Multilingual: German, English, Czech and French
  • Operations: Start, Stop, Cycle time, Read data, Delete data.

Chart display: Various measuring sequences in one diagram, real-time axis, zoom function, measurement points, legend and measurement cursor displayed or hidden, add comments.
System requirements: 1 GHz CPU, 1 GB RAM, 100 MB HDD, Windows 7 SP 1 (32 or 64 Bit), interface converter for GMH 3000: GRS 3100, GRS 3105 or USB 3100 N and for GMH 5000: USB 5100 (executable from Windows XP, not executable with Windows RT, ARM or Intel Itanium based Windows systems).

Specifications
  • Description:
    Windows software, GSOFT 40K
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.