ContactUsLinkComponent
VWR® RM 10, Moisture Tester
VWR® RM 10, Moisture Tester
Catalog # 620-1956
Supplier:  VWR Collection
VWR® RM 10, Moisture Tester
Catalog # 620-1956
Supplier:  VWR Collection
Specifications
  • Model:
    RM 10
  • Description:
    Moisture tester
  • Pk:
    1
Specifications
About this item
The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or wash floor down to 4 cm.
  • Easy and fast moisture rating, non destructive measurement
  • Displayed values by „digit“ are relative so that values can be compared
  • To check moisture on surfaces – No injection into the measuring object required
Suitable for high-speed determination of moisture i.e. estate agents, property management, house owners, architects, building experts, building contractors.
Caution: This is an indicator for fast estimation – Does not replace precision instruments like the RM 20.