ContactUsLinkComponent
Adapter for safety exhaust filters
Adapter for safety exhaust filters
  JOURJR-S-40004
Adapter for safety exhaust filters
  JOURJR-S-40004
  • Description:
    Adapter, connects safety exhaust filter to SCAT® waste caps and A.I.T. smart waste caps
  • Seq:
    03
  • Pk:
    1