Reagent for sulphate determination using Lovibond® MD photometers
Catalog # LOVI515450BT
Supplier: Lovibond
CAS Number:
Specifications
- Chemical method:Barium sulphate (turbidity)
- Pk:100 Tab.
- Description:Sulphate tablet
- Environmentally Preferable:
- For unit:MD600, MD610, MultiDirect and Spectrodirect
- Range:5 - 100 mg/l SO₄