Orders LinkContactUsLinkComponent
Sorbents, LiChrosorb®
Sorbents, LiChrosorb®
Supplier:   Merck
Sorbents, LiChrosorb®
Supplier:  Merck
Certificates
Sorbents, LiChrosorb®
Sorbents, LiChrosorb®
5 Options Available Below
Product Details & Documents
LiChrosorb® is an irregular shaped silica sorbent. LiChrosorb® packing materials offer the compete programme of non-polar derivatives (RP-8, RP-18, RP-select B) and polar derivatives (Si 60 and Si 100).
Product Family Options
Product Information
  • Particle Size
    Pk
    Availability
    Price
  • 1.09309.0010
    LiChrosorb® Si 100
    LiChrosorb® Si 100
    10 µm
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 235,00 €
    235,00 €
    1 * 10 g
       235,00 €
  • 1.09318.0010
    LiChrosorb® RP-8
    LiChrosorb® RP-8
    10 µm
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 020,00 €
    1 020,00 €
    1 * 10 g
       1 020,00 €
  • 1.09332.0010
    LiChrosorb® RP-8
    LiChrosorb® RP-8
    5 µm
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 110,00 €
    1 110,00 €
    1 * 10 g
       1 110,00 €
  • 1.09333.0010
    LiChrosorb® RP-18
    LiChrosorb® RP-18
    5 µm
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 110,00 €
    1 110,00 €
    1 * 10 g
       1 110,00 €
  • 1.09334.0010
    LiChrosorb® RP-18
    LiChrosorb® RP-18
    10 µm
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 050,00 €
    1 050,00 €
    1 * 10 g
       1 050,00 €