Multi-parameter meter (pH/conductivity), handheld, 430
Catalog # 662-1175
Supplier: Cole-Parmer
Specifications
- Temperature range:–9,9...+99,9 °C
- Description:Handheld pH/conductivity meter 430
- Model:430
- Temperature accuracy:±0,5 °C
- Temperature resolution:0,1 °C
- Conductivity accuracy:±0,5% ±2 digits
- Conductivity calibration:Automatic standard recognition: 10 µS, 84 µS, 1413 µS, 12,88 mS (with manual override)
- Conductivity cell constant:0,01 - 19,99 cm⁻¹
- Conductivity range:5 auto-selected from 0 to 199,9 mS/cm
- Conductivity resolution:0,01 µS - 0,1 mS
- Connections:Mini-DIN
- TDS accuracy:±0,5% ±2 digits
- TDS range:5 auto-selected from 0 to 199,9 g/l
- TDS resolution:0,01 / 0,1
- Temperature compensation:ATC and manual, 0 to +99,9 °C
- Weight:0,250 kg
- W×D×H:75×35×175 mm
- pH accuracy:±0,02
- pH calibration:1 or 2-point
- pH range:0,00...+14,00
- pH resolution:0,01
- Pk:1
Specifications
About this item
The easy-to-use 430 is designed to readily withstand the rigours of field work, while reliably measuring pH and conductivity.
- Simultaneous display of pH, conductivity and temperature
- Automatic or manual calibration
- Power on self check routine
- Auto-selecting ranges for both TDS and conductivity modes
Ordering information: Supplied with combined pH/conductivity probe, buffer sachets, batteries (fitted) and free carry case.