Orders LinkContactUsLinkComponent
Sample bottles, glass
Sample bottles, glass
Supplier:   BURKLE
Sample bottles, glass
Supplier:  BURKLE
Certificates
Sample bottles, glass
Sample bottles, glass
3 Options Available Below
Product Details & Documents
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Product Family Options
Product Information
  • Capacity
    Thread
    Availability
    Price
  • 300-0054
    Sample bottles
    Sample bottles
    100 ml
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,10 €
    14,10 €
    1 * 1 ST
       14,10 €
  • 300-0055
    Sample bottles
    Sample bottles
    250 ml
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,10 €
    24,10 €
    1 * 1 ST
       24,10 €
  • BURK5314-0100
    Sample bottles
    Sample bottles
    1000 ml
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,50 €
    26,50 €
    1 * 1 ST
       26,50 €