Connection stopper
Connection stopper for distribution heads with connection pipe Ø 32,7 mm
Catalog # 458-2357
Supplier: GERHARDT
Jump to:
Product Details & Documents
This connection stopper is made from viton and is suitable for distribution heads with connection pipe Ø 32,7 mm.
Specifications
- Description:Connection stopper for distribution heads with connection pipe Ø 32,7 mm