Sample dippers, DispoDipper
Supplier: BURKLE
Jump to:
About this item
The DispoDipper are designed for the sampling and subsequent transport of samples.
- Production and packaging according to cleanroom class 7
- Prevents cross-contamination or sample impurities
- Optionally sterilised by gamma rays (SteriPlast®)
Suitable for applications in food, animal feed and pharmaceutical industries.
: EU food requirements and FDA regulations.
: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
6 Options Available Below
Product Details & Documents
The DispoDipper are designed for the sampling and subsequent transport of samples.
- Production and packaging according to cleanroom class 7
- Prevents cross-contamination or sample impurities
- Optionally sterilised by gamma rays (SteriPlast®)
Suitable for applications in food, animal feed and pharmaceutical industries.
: EU food requirements and FDA regulations.
: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Product Family Options
Product Information
- DésignationMatièreLongueurØStérileCapacitéAvailabilityPrice