ContactUsLinkComponent
Microarray scanner, GenePix® 4000B
Microarray scanner, GenePix® 4000B
Catalog # MLDVGENEPIX4000B-U
Supplier:  Molecular Devices
undefined
Microarray scanner, GenePix® 4000B
Catalog # MLDVGENEPIX4000B-U
Supplier:  Molecular Devices
Specifications
  • Description:
    Microarray scanner, GenePix® 4000B
  • Pk:
    1
Specifications
About this item
The GenePix® 4000B microarray scanner is a benchmark for quality, reliability and ease-of-use in microarray scanning technology. Coupled with GenePix® Pro microarray image analysis software and Acuity® microarray informatics software, the GenePix system sets the highest standards in the acquisition and analysis of data from all types of arrays, including nucleic acids, proteins, tissues, and cells.
  • Simultaneous dual-laser scanning
  • Reliability and ease-of-use
  • Automated workflow
With automated photomultiplier (PMT) balancing, the fully integrated GenePix Pro acquisition and analysis software controls the GenePix 4000B microarray scanner and can automatically evaluate the required PMT gain values for fast and easy optimisation of signal intensity and channel balance.