Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™
Certificates
Buffer solutions, colour coded, Fluka™

20 Options Available Below

Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Product Family Options

Product Information

  • pH value
    Colour
    Reference temperature
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • 4,00
    Red
    20 °C
    Plastic bottle
    0,5 L
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 42,40 €
    42,40 €
    1 * 500 mL
       42,40 €

  • 4,00
    Red
    20 °C
    Plastic bottle
    6×0,5 L
    3.000 mL
    Log in for availability
    Log in for availability
    1 * 3.000 mL
    1 * 3.000 mL - 224,00 €
    224,00 €
    1 * 3.000 mL
       224,00 €

  • 4,00
    Red
    20 °C
    Plastic bottle
    1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 86,00 €
    86,00 €
    1 * 1 L
       86,00 €

  • 4,00
    Red
    20 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 451,00 €
    451,00 €
    1 * 6 L
       451,00 €

  • 4,00
    Red
    20 °C
    VOLPAK container
    5 L
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 130,00 €
    130,00 €
    1 * 5 L
       130,00 €

  • 4,00
    Red
    20 °C
    VOLPAK container
    10 L
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 310,00 €
    310,00 €
    1 * 10 L
       310,00 €

  • 7,00
    Green
    20 °C
    Plastic bottle
    0,5 L
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,30 €
    41,30 €
    1 * 500 mL
       41,30 €

  • 7,00
    Green
    20 °C
    Plastic bottle
    6×0,5 L
    3.000 mL
    Log in for availability
    Log in for availability
    1 * 3.000 mL
    1 * 3.000 mL - 216,00 €
    216,00 €
    1 * 3.000 mL
       216,00 €

  • 7,00
    Green
    20 °C
    Plastic bottle
    1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 86,80 €
    86,80 €
    1 * 1 L
       86,80 €

  • 7,00
    Green
    20 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 456,00 €
    456,00 €
    1 * 6 L
       456,00 €

  • 7,00
    Green
    20 °C
    VOLPAK container
    5 L
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 131,00 €
    131,00 €
    1 * 5 L
       131,00 €

  • 7,00
    Green
    20 °C
    VOLPAK container
    10 L
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 311,00 €
    311,00 €
    1 * 10 L
       311,00 €

  • 9,00
    Blue
    20 °C
    Plastic bottle
    1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 86,40 €
    86,40 €
    1 * 1 L
       86,40 €

  • 9,00
    Blue
    20 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 451,00 €
    451,00 €
    1 * 6 L
       451,00 €

  • 9,00
    Blue
    20 °C
    VOLPAK container
    5 L
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 136,00 €
    136,00 €
    1 * 5 L
       136,00 €

  • 10,00
    Violet
    20 °C
    Plastic bottle
    0,5 L
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,30 €
    39,30 €
    1 * 500 mL
       39,30 €

  • 10,00
    Violet
    20 °C
    Plastic bottle
    6×0,5 L
    3.000 mL
    Log in for availability
    Log in for availability
    1 * 3.000 mL
    1 * 3.000 mL - 205,00 €
    205,00 €
    1 * 3.000 mL
       205,00 €

  • 10,00
    Violet
    20 °C
    Plastic bottle
    1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 78,90 €
    78,90 €
    1 * 1 L
       78,90 €

  • 10,00
    Violet
    20 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 420,00 €
    420,00 €
    1 * 6 L
       420,00 €

  • 10,00
    Violet
    20 °C
    VOLPAK container
    10 L
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 262,00 €
    262,00 €
    1 * 10 L
       262,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.