Orders LinkContactUsLinkComponent
Pasteur pipettes
Supplier:   Greiner Bio-One
Pasteur pipettes
Supplier:  Greiner Bio-One
Certificates
Pasteur pipettes
4 Options Available Below
Product Details & Documents
These pipettes are made from PE.
Product Family Options
Product Information
  • Capacity
    Drop vol.
    Length
    Sterile
    Packed
    Availability
    Price
  • Graduated
    1 ml
    35 µl
    150 mm
    -
    500/inner pack, 3000/carton
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 177,00 €
    177,00 €
    1 * 3.000 ST
       177,00 €
  • Graduated
    1 ml
    35 µl
    150 mm
    +
    10/inner pack, 500/carton
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 86,30 €
    86,30 €
    1 * 500 ST
       86,30 €
  • Graduated
    1 ml
    35 µl
    150 mm
    +
    Individually wrapped, 500/carton
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 93,80 €
    93,80 €
    1 * 500 ST
       93,80 €
  • Long
    6 ml
    48 µl
    230 mm
    -
    400/inner pack, 2400/carton
    Log in for availability
    Log in for availability
    1 * 2.400 ST
    1 * 2.400 ST - 470,00 €
    470,00 €
    1 * 2.400 ST
       470,00 €