Protamine sulphate (from salmon sperm), ≥100 USP u/mg, white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9009-65-8
MDL Number:MFCD00132091
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Source:Salmon Sperm
- Loss on Drying:≤5%
- Sulfate:16-22%
- Nitrogen:22.5-25.5%
- Activity:≥100 USP u/mg
Specification Test Results
Frequently Bought Together