Order Entry
Belgium
ContactUsLinkComponent
 
Single element petroleum atandard, calcium in hydrocarbon oil, CRM, VHG
  PROOVHGOCA5000-50G
 :  
Single element petroleum atandard, calcium in hydrocarbon oil, CRM, VHG
  PROOVHGOCA5000-50G
 :  

 

  • Concentration:
    5000 µg/g
  • Matrix:
    oil
  • Pk:
    50 g
  • Element:
    Calcium
  • Symbol:
    Ca

 

 

This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.

 : Traceable to NIST, SRM.