Orders LinkContactUsLinkComponent
Pipe blind lockouts
Pipe blind lockouts
Supplier:   Brady
Pipe blind lockouts
Supplier:  Brady
Certificates
Pipe blind lockouts
Pipe blind lockouts
3 Options Available Below
Product Details & Documents
These devices can be applied to the widest range of pipes to easily and temporarily avoid the release of gas, steam and hazardous liquids while pipe maintenance is still ongoing.
  • Adjustable locking arm accommodates both standard and recessed bolt positions
  • Secure 1 or 2 bolts for extra security
  • Heavy duty construction withstands harsh environments
  • UL approved for use in explosive atmospheres
Min. shackle clearance: 19,05 mm
Max. shackle diameter: 7,00 mm
Product Family Options
Product Information
  • Type
    Colour
    W×D×H
    Availability
    Price
  • Pipe blind lockout, small (hand wheel diameter 13 - 76 mm)
    Red
    374,65×177,8×73,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • Pipe blind lockout, medium (hand wheel diameter 76 - 355 mm)
    Red
    374,65×215,9×73,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 119,00 €
    119,00 €
    1 * 1 ST
       119,00 €
  • Pipe blind lockout, large (hand wheel diameter 355 - 1219 mm)
    Red
    628,65×247,65×92,08 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €