Order Entry
Belgium
ContactUsLinkComponent
Dasatinib monohydrate
Dasatinib monohydrate
  ACRO462320010
 :  
undefined
Dasatinib monohydrate
  ACRO462320010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

 

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    2 to 4 %
  • HPLC:
    ≥98.5 %
  • Residue after ignition:
    ≤0.2 %

 

 

 : Dasatinib hydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide monohydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrate
 :
 :506.02 g/mol
 :Freezer
 :2811
 :6.1,III
 :GHS06
 :GHS08
 :863127-77-9
 :MFCD11046566
 :C₂₂H₂₆ClN₇O₂S·H₂O