Order Entry
Belgium
ContactUsLinkComponent
N-Ethyldiisopropylamine 99.5+%, AcroSeal®
Flammable
Corrosive
Toxic
N-Ethyldiisopropylamine 99.5+%, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Toxic
N-Ethyldiisopropylamine 99.5+%, AcroSeal®
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4130 to 1.4150 (20°C, nm)
  • Water:
    ≤0.3 %
  • GC:
    ≥99.50 %
  • Color scale:
    ≤50 APHA

Specification Test Results

Related Information

Frequently Bought Together