Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Anthralin 97%

Irritant
Anthralin 97%
Certificates
Anthralin 97%

3 Options Available Below

Product Details & Documents
Synonyms:  1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
  • CAS Number:1143-38-0
  • Formula:C₁₄H₁₀O₃
  • MDL Number:MFCD00053409
  • Molecular Weight:226.23 g/mol
  • Melting Pt:178…181 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to red Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    178 to 182 °C
  • HPLC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 28,40 €
    28,40 €
    1 * 1 g
       28,40 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 98,30 €
    98,30 €
    1 * 5 g
       98,30 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 379,00 €
    379,00 €
    1 * 25 g
       379,00 €