Order Entry
Belgium
ContactUsLinkComponent
L(+)-Asparagine, anhydrous ≥99%, white powder cell culture reagent
L(+)-Asparagine, anhydrous ≥99%, white powder cell culture reagent
Supplier:  MP Biomedicals
Certificates
undefined
L(+)-Asparagine, anhydrous ≥99%, white powder cell culture reagent
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Loss on Drying:
    ≤1.0%
  • Optical Rotation:
    +25° to +36° (c=10, 1N AM56HCl)

Specification Test Results

Frequently Bought Together