Order Entry
Belgium
ContactUsLinkComponent
Potassium carbonate, anhydrous 98%, extra pure
Irritant
Potassium carbonate, anhydrous 98%, extra pure
Certificates
undefined
Irritant
Potassium carbonate, anhydrous 98%, extra pure

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤2.0 %
  • Titration with HClO4:
    ≥97.5 %
  • Heavy metals (as Pb):
    ≤0.005 %

Specification Test Results

Frequently Bought Together