Order Entry
Belgium
ContactUsLinkComponent
 
Sodium acetate trihydrate for HPLC
 
undefined
Sodium acetate trihydrate for HPLC
 

 

  • Appearance:
    White Crystals
  • UV:
    (1 M in water)
  • pH:
    7.5 to 9.2
  • Titration with HClO4:
    ≥98.5 %
  • Insoluble matter:
    ≤0.0005 %

 

 

Frequently Bought Together