Ammonium cerium(IV) nitrate, pure
:16774-21-3
:
:(NH₄)₂Ce(NO₃)₆
:MFCD00151121
:548.22 g/mol
:107…108 °C
:3085
:5.1,II
:GHS07
:GHS05
:GHS03
:Ambient
- Appearance:Orange Crystalline powder or crystals
- Loss on drying:≤2 %
- Titration:≥98.5 %
Frequently Bought Together