Order Entry
Belgium
ContactUsLinkComponent
Ammonium iron(II) sulphate hexahydrate 98%, extra pure
Ammonium iron(II) sulphate hexahydrate 98%, extra pure
Catalog # ACRO193250050
CAS Number:  
undefined
Ammonium iron(II) sulphate hexahydrate 98%, extra pure
Catalog # ACRO193250050
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Seq:
    4114
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Green to blue-green to blue to dark blue Powder
  • pH:
    3 to 5
  • Titration with KMnO4:
    ≥97.5 %
  • Insoluble matter:
    ≤0.05 % (in dilute H2SO4)

Specification Test Results

About this item

Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
 :
 :392.14 g/mol
Melting Pt:100 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:7783-85-9
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530