Orders LinkContactUsLinkComponent
Chromatography columns with filter plates
Chromatography columns with filter plates
Certificates
Chromatography columns with filter plates
6 Options Available Below
Product Details & Documents
These chromatography columns feature a top sleeve with PTFE tap and are made from borosilicate glass 3.3.
Product Family Options
Product Information
  • Ø int.
    Length
    Socket size
    Availability
    Price
  • NEUB1202-14-015030
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    15,00 mm
    300 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 210,00 €
    210,00 €
    1 * 1 ST
       210,00 €
  • NEUB1202290200200
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    20,00 mm
    200 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €
  • NEUB1202-290300300
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    30,00 mm
    300 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 209,00 €
    209,00 €
    1 * 1 ST
       209,00 €
  • NEUB1202-290400400
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    40,00 mm
    400 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 322,00 €
    322,00 €
    1 * 1 ST
       322,00 €
  • NEUB1202450600600
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    60,00 mm
    600 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 451,00 €
    451,00 €
    1 * 1 ST
       451,00 €
  • NEUB1202-290800400
    Chromatography column with PTFE tap
    Chromatography column with PTFE tap
    80,00 mm
    400 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 672,00 €
    672,00 €
    1 * 1 ST
       672,00 €