2-Amino-6-(trifluoromethoxy)benzothiazole ≥97.0% (by HPLC), Calbiochem®
Catalog # 557324-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Aluminium drum
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Form:White to off-white solid
- Purity:≥97% by HPLC
- Contaminants:Heavy metals: ≤20 ppm
- Storage:+2 °C to +8 °C
- Stability:Stable for up to 3 months at –20 °C
Specification Test Results
About this item
:
:234.20 g/mol
Storage Temperature:Refrigerator
CAS Number:1744-22-5
Formula:C₈H₅F₃N₂OS
MDL Number:MFCD00210213