Order Entry
Belgium
ContactUsLinkComponent
Silicon dioxide ~99% 0.5-10 μm (∼ 80% 1-5 μm), Fluka™
Health
Silicon dioxide ~99% 0.5-10 μm (∼ 80% 1-5 μm), Fluka™
Certificates
Health
Silicon dioxide ~99% 0.5-10 μm (∼ 80% 1-5 μm), Fluka™
Certificates

Specification Test Results

  • Particle Size:
    0.5-10 μm (approx. 80% between 1-5 μm)
  • Assay:
    Min. 98 %
  • Grain size < 8 micron:
    Min. 97 %

Specification Test Results

Related Information

Frequently Bought Together