Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 35 - 38%, AR®, Macron Fine Chemicals™
Corrosive
Irritant
Hydrochloric acid 35 - 38%, AR®, Macron Fine Chemicals™
Certificates
Corrosive
Irritant
Hydrochloric acid 35 - 38%, AR®, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Appearance:
    Clear, Colourless to Yellow Liquid
  • Assay:
    35 - 38%
  • Iron (Fe):
    Max. 0.0001%
  • Free chlorine (Cl₂):
    Max. 0.001%
  • Heavy metals (as Pb):
    Max. 0.0001%
  • Residue after Ignition:
    Max. 0.001%
  • Sulphate (SO₄):
    Max. 0.001%

Specification Test Results

Related Information

Frequently Bought Together