Combined density and refractometry standards
Supplier: Mettler - Toledo
Jump to:
About this item
These combined reference standards are intended to reduce uncertainty, eliminate time-consuming and error prone sample transfer and facilitate fully automatic performance verification.
2 Options Available Below
Product Details & Documents
These combined reference standards are intended to reduce uncertainty, eliminate time-consuming and error prone sample transfer and facilitate fully automatic performance verification.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Refractive indexPkAvailabilityPrice
- METT513380132,4-dichlorotoluene, 9 ml
- METT513380141-bromonaphthalene, 9 ml