Order Entry
Belgium
ContactUsLinkComponent
Plasmid isolation kits, NucleoSpin® Plasmid
Plasmid isolation kits, NucleoSpin® Plasmid
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Plasmid isolation kits, NucleoSpin® Plasmid
Supplier:  MACHEREY-NAGEL
Certificates

Specifications

  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample size:
    2 − 10 ml
  • Description:
    NucleoSpin® Plasmid

Specifications

Related Information

Frequently Bought Together