ContactUsLinkComponent
Deep well plates, 24-well no-bottom plates
Deep well plates, 24-well no-bottom plates
Catalog # GREI662000-06_28
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Deep well plates, 24-well no-bottom plates
Catalog # GREI662000-06_28
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Colour:
    Black
  • Description:
    Deep well plates, 24-well no bottom plates
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    28
Specifications
About this item
Plates are made from virgin polystyrene resin and molded without the bottom of the plate. They feature chimney-well design to help prevent cross-contamination.