Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ion chromatography standards, fluoride

Ion chromatography standards, fluoride

Fluoride standard solution, 1000 mg/l, NaF in H₂O, Plastic canister, natural, 5 L
Catalog # KRAF22076.5000
Supplier:   Bernd Kraft
Ion chromatography standards, fluoride
Catalog # KRAF22076.5000
Supplier:  Bernd Kraft
CAS Number:  
Ion chromatography standards, fluoride
Ion chromatography standards, fluoride

Fluoride standard solution, 1000 mg/l, NaF in H₂O, Plastic canister, natural, 5 L

CAS Number:  
Price
144,00 €
Log in for availability
1 * 5 L
Product Details & Documents

NaF in water standard solution having shelf life of 1092 days.

Specifications

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic canister, natural
  • Pk:
    5 L
  • Description:
    Fluoride standard solution
  • Contents:
    NaF in H₂O

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.