Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 6 mol/l (6 N) in aqueous solution for biopharmaceutical production
Corrosive
Sodium hydroxide 6 mol/l (6 N) in aqueous solution for biopharmaceutical production
Catalog # 85522.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 6 mol/l (6 N) in aqueous solution for biopharmaceutical production
Catalog # 85522.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    5.9 --> 6.1 mol/l
  • Appearance:
    Passes test
  • CO3 (as Na2CO3):
    Max. 1 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Fe (Iron):
    Max. 3 ppm
  • Residual metal catalysts:
    Unlikely to be present
  • Residual solvents (ICH Q3C):
    Excl.by manufact. process
  • Endotoxin:
    Max. 2.0 EU/ml
  • Filtered through 0.2 μm:
    Confirmed

Specification Test Results

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548